Product Details

SNP ID
rs117359246
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:240572557 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGTGAAGTGCACCTACTCTGCCGT[C/T]GTCAAGGTCAGGGGCCGCCAGCTGC
Phenotype
MIM: 605287
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DUSP28 PubMed Links

Gene Details

Gene
DUSP28
Gene Name
dual specificity phosphatase 28
There are no transcripts associated with this gene.

Gene
RNPEPL1
Gene Name
arginyl aminopeptidase like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018226.4 663 Silent Mutation GTC,GTT V221V NP_060696.4
XM_005247036.4 663 Silent Mutation GTC,GTT V221V XP_005247093.1

View Full Product Details