Product Details

SNP ID
rs116809963
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:116712606 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCAAAGCAACAACTGAGGTGGCG[C/T]ATGAAAACTGTCTACAAGCATTTGA
Phenotype
MIM: 616057
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TUSC7 PubMed Links

Gene Details

Gene
TUSC7
Gene Name
tumor suppressor candidate 7 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details