Product Details

SNP ID
rs112071257
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:119642720 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATAGGGGCAGGTGTACCTTCGAAT[C/T]ATTTCATACTAGAATATTAGGGCAA
Phenotype
MIM: 605823
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POPDC2 PubMed Links

Gene Details

Gene
POPDC2
Gene Name
popeye domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308333.1 Intron NP_001295262.1
NM_022135.3 Intron NP_071418.2
XM_005247698.3 Intron XP_005247755.1

View Full Product Details