Product Details

SNP ID
rs113238208
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:120687650 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATTTACCTTAGATTATTAAAAAC[C/T]ATTTTTTCTTAGAAAACTATAATTC
Phenotype
MIM: 607474
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HGD PubMed Links

Gene Details

Gene
HGD
Gene Name
homogentisate 1,2-dioxygenase
There are no transcripts associated with this gene.

Gene
RABL3
Gene Name
RAB, member of RAS oncogene family like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173825.3 2826 UTR 3 NP_776186.2
XM_006713595.2 2826 UTR 3 XP_006713658.1
XM_017006227.1 2826 UTR 3 XP_016861716.1
XM_017006228.1 2826 UTR 3 XP_016861717.1

View Full Product Details