Product Details

SNP ID
rs115356519
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:121666081 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTATGGAGGTCAGTCTGTCAATG[C/T]AGGCTATGCTGAGGAAGGATGAGAA
Phenotype
MIM: 602500 MIM: 601306
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GOLGB1 PubMed Links

Gene Details

Gene
GOLGB1
Gene Name
golgin B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256486.1 Intron NP_001243415.1
NM_001256487.1 Intron NP_001243416.1
NM_001256488.1 Intron NP_001243417.1
NM_004487.4 Intron NP_004478.3
XM_005247371.4 Intron XP_005247428.1
XM_005247372.4 Intron XP_005247429.1
XM_005247373.1 Intron XP_005247430.1
XM_006713587.1 Intron XP_006713650.1
XM_006713588.1 Intron XP_006713651.1
XM_006713589.1 Intron XP_006713652.1
XM_006713590.1 Intron XP_006713653.1
XM_006713591.1 Intron XP_006713654.1
XM_011512699.2 Intron XP_011511001.1
XM_017006189.1 Intron XP_016861678.1
XM_017006190.1 Intron XP_016861679.1
XM_017006191.1 Intron XP_016861680.1
XM_017006192.1 Intron XP_016861681.1
XM_017006193.1 Intron XP_016861682.1
XM_017006194.1 Intron XP_016861683.1
XM_017006195.1 Intron XP_016861684.1
Gene
HCLS1
Gene Name
hematopoietic cell-specific Lyn substrate 1
There are no transcripts associated with this gene.

View Full Product Details