Product Details

SNP ID
rs115407410
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:122384273 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGAGTAGCAAGTGGCCATGGGGA[A/G]CCTCAGCGGTCTGCGCCTGGCAGCA
Phenotype
MIM: 608017
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC58 PubMed Links

Gene Details

Gene
CCDC58
Gene Name
coiled-coil domain containing 58
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017928.3 98 Intron NP_001017928.1
NM_001308326.1 98 Intron NP_001295255.1
XM_011512410.2 98 Intron XP_011510712.1
XM_017005697.1 98 Intron XP_016861186.1
Gene
FAM162A
Gene Name
family with sequence similarity 162 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014367.3 98 Missense Mutation AAC,AGC N3S NP_055182.3

View Full Product Details