Product Details

SNP ID
rs115815057
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:123613892 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCTAAATGAAATCTAGCTGCACT[A/T]GTATCTTAAGGTGCCAACTAACATA
Phenotype
MIM: 600922
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MYLK PubMed Links

Gene Details

Gene
MYLK
Gene Name
myosin light chain kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321309.1 2557 UTR 3 NP_001308238.1
NM_053025.3 2557 UTR 3 NP_444253.3
NM_053026.3 2557 UTR 3 NP_444254.3
NM_053027.3 2557 UTR 3 NP_444255.3
NM_053028.3 2557 UTR 3 NP_444256.3
XM_011512860.2 2557 Intron XP_011511162.1
XM_011512861.2 2557 Intron XP_011511163.1
XM_017006469.1 2557 UTR 3 XP_016861958.1
XM_017006470.1 2557 UTR 3 XP_016861959.1
XM_017006471.1 2557 UTR 3 XP_016861960.1
XM_017006472.1 2557 UTR 3 XP_016861961.1
XM_017006473.1 2557 UTR 3 XP_016861962.1
Gene
MYLK-AS1
Gene Name
MYLK antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details