Product Details
- SNP ID
-
rs113329897
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:123614238 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTGGCATCGTCATCCCCGCAAACA[C/T]CACTAATAATTAAAGAGCAGTTCCC
- Phenotype
-
MIM: 600922
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MYLK
PubMed Links
Gene Details
- Gene
- MYLK
- Gene Name
- myosin light chain kinase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001321309.1 |
2211 |
Missense Mutation |
GAT,GGT |
D1695G |
NP_001308238.1 |
NM_053025.3 |
2211 |
Missense Mutation |
GAT,GGT |
D1871G |
NP_444253.3 |
NM_053026.3 |
2211 |
Missense Mutation |
GAT,GGT |
D1802G |
NP_444254.3 |
NM_053027.3 |
2211 |
Missense Mutation |
GAT,GGT |
D1820G |
NP_444255.3 |
NM_053028.3 |
2211 |
Missense Mutation |
GAT,GGT |
D1751G |
NP_444256.3 |
XM_011512860.2 |
2211 |
Missense Mutation |
GAT,GGT |
D1870G |
XP_011511162.1 |
XM_011512861.2 |
2211 |
Missense Mutation |
GAT,GGT |
D1803G |
XP_011511163.1 |
XM_017006469.1 |
2211 |
Missense Mutation |
GAT,GGT |
D948G |
XP_016861958.1 |
XM_017006470.1 |
2211 |
Missense Mutation |
GAT,GGT |
D670G |
XP_016861959.1 |
XM_017006471.1 |
2211 |
Missense Mutation |
GAT,GGT |
D671G |
XP_016861960.1 |
XM_017006472.1 |
2211 |
Missense Mutation |
GAT,GGT |
D111G |
XP_016861961.1 |
XM_017006473.1 |
2211 |
Missense Mutation |
GAT,GGT |
D110G |
XP_016861962.1 |
- Gene
- MYLK-AS1
- Gene Name
- MYLK antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details