Product Details

SNP ID
rs116461370
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:129060185 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGTACTGCAAATGTTAAAACCA[C/T]AGGCCAAAATTCTAGCTAGAGATTC
Phenotype
MIM: 173515
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GP9 PubMed Links

Gene Details

Gene
GP9
Gene Name
glycoprotein IX platelet
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000174.4 Intron NP_000165.1
XM_005247374.3 Intron XP_005247431.1
XM_011512701.1 Intron XP_011511003.1
XM_011512702.1 Intron XP_011511004.1

View Full Product Details