Product Details

SNP ID
rs112843206
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:98354020 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAATTTATGTAGAGCGTCGTCTTC[C/T]CACACCAATGTACATCTTTCTGGGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR5K4 PubMed Links

Gene Details

Gene
OR5K4
Gene Name
olfactory receptor family 5 subfamily K member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005517.1 167 Missense Mutation CCC,CTC P56L NP_001005517.1

View Full Product Details