Product Details
- SNP ID
-
rs112292133
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:139561222 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGTGAATGTTGTGCTGGTGCATCC[C/G]TAGGATGGGAGATTCTGCGATGTAA
- Phenotype
-
MIM: 608702
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LOC100507291
PubMed Links
Gene Details
- Gene
- LOC100507291
- Gene Name
- uncharacterized LOC100507291
There are no transcripts associated with this gene.
- Gene
- NMNAT3
- Gene Name
- nicotinamide nucleotide adenylyltransferase 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001200047.2 |
1044 |
Missense Mutation |
CGG,GGG |
R94G |
NP_001186976.1 |
NM_001320510.1 |
1044 |
Missense Mutation |
CGG,GGG |
R277G |
NP_001307439.1 |
NM_001320511.1 |
1044 |
Missense Mutation |
CGG,GGG |
R183G |
NP_001307440.1 |
NM_001320512.1 |
1044 |
Missense Mutation |
CGG,GGG |
R183G |
NP_001307441.1 |
NM_001320513.1 |
1044 |
Missense Mutation |
CGG,GGG |
R73G |
NP_001307442.1 |
NM_178177.4 |
1044 |
Missense Mutation |
CGG,GGG |
R146G |
NP_835471.1 |
XM_011512790.1 |
1044 |
Missense Mutation |
CGG,GGG |
R146G |
XP_011511092.1 |
XM_011512791.2 |
1044 |
Missense Mutation |
CGG,GGG |
R146G |
XP_011511093.1 |
XM_017006328.1 |
1044 |
Missense Mutation |
CGG,GGG |
R277G |
XP_016861817.1 |
XM_017006329.1 |
1044 |
Missense Mutation |
CGG,GGG |
R277G |
XP_016861818.1 |
XM_017006330.1 |
1044 |
Missense Mutation |
CGG,GGG |
R277G |
XP_016861819.1 |
XM_017006331.1 |
1044 |
Missense Mutation |
CGG,GGG |
R277G |
XP_016861820.1 |
XM_017006332.1 |
1044 |
Missense Mutation |
CGG,GGG |
R277G |
XP_016861821.1 |
XM_017006333.1 |
1044 |
Missense Mutation |
CGG,GGG |
R277G |
XP_016861822.1 |
XM_017006334.1 |
1044 |
Missense Mutation |
CGG,GGG |
R277G |
XP_016861823.1 |
XM_017006335.1 |
1044 |
Missense Mutation |
CGG,GGG |
R183G |
XP_016861824.1 |
XM_017006336.1 |
1044 |
Missense Mutation |
CGG,GGG |
R183G |
XP_016861825.1 |
XM_017006337.1 |
1044 |
Missense Mutation |
CGG,GGG |
R183G |
XP_016861826.1 |
XM_017006338.1 |
1044 |
Missense Mutation |
CGG,GGG |
R146G |
XP_016861827.1 |
XM_017006339.1 |
1044 |
UTR 3 |
|
|
XP_016861828.1 |
XM_017006340.1 |
1044 |
UTR 3 |
|
|
XP_016861829.1 |
XM_017006341.1 |
1044 |
UTR 3 |
|
|
XP_016861830.1 |
XM_017006342.1 |
1044 |
UTR 3 |
|
|
XP_016861831.1 |
XM_017006343.1 |
1044 |
UTR 3 |
|
|
XP_016861832.1 |
XM_017006344.1 |
1044 |
UTR 3 |
|
|
XP_016861833.1 |
View Full Product Details