Product Details

SNP ID
rs121434460
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:146071407 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAGTTGGGACATTTTCATAACCA[C/A]CAGATATACGGCTATCCTAGAAACA
Phenotype
MIM: 601865
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
PLOD2 PubMed Links

Gene Details

Gene
PLOD2
Gene Name
procollagen-lysine,2-oxoglutarate 5-dioxygenase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000935.2 1928 Missense Mutation GGT,GTT G601V NP_000926.2
NM_182943.2 1928 Missense Mutation GGT,GTT G622V NP_891988.1
XM_005247535.3 1928 Missense Mutation GGT,GTT G530V XP_005247592.1
XM_017006625.1 1928 Missense Mutation GGT,GTT G530V XP_016862114.1

View Full Product Details