Product Details

SNP ID
rs115817835
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:149372163 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCTAACAGGCATCTGAATTCTTA[C/T]AAACAAAAATAAAAATGCAGAACTG
Phenotype
MIM: 191155
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TM4SF1 PubMed Links

Gene Details

Gene
TM4SF1
Gene Name
transmembrane 4 L six family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014220.2 Intron NP_055035.1
XM_017006385.1 Intron XP_016861874.1
Gene
TM4SF1-AS1
Gene Name
TM4SF1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details