Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_053002.5 | 1247 | Intron | NP_443728.3 | ||
XM_006713487.3 | 1247 | Intron | XP_006713550.1 | ||
XM_011512390.2 | 1247 | Intron | XP_011510692.1 | ||
XM_011512394.2 | 1247 | Intron | XP_011510696.1 | ||
XM_011512399.2 | 1247 | Intron | XP_011510701.1 | ||
XM_017005676.1 | 1247 | Intron | XP_016861165.1 | ||
XM_017005677.1 | 1247 | Intron | XP_016861166.1 | ||
XM_017005678.1 | 1247 | Intron | XP_016861167.1 | ||
XM_017005679.1 | 1247 | Intron | XP_016861168.1 | ||
XM_017005680.1 | 1247 | Intron | XP_016861169.1 | ||
XM_017005681.1 | 1247 | Intron | XP_016861170.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001081455.1 | 1247 | Missense Mutation | CCA,CTA | P320L | NP_001074924.1 |
NM_014879.3 | 1247 | Missense Mutation | CCA,CTA | P320L | NP_055694.3 |
XM_005247922.3 | 1247 | Missense Mutation | CCA,CTA | P320L | XP_005247979.1 |
XM_005247923.3 | 1247 | Missense Mutation | CCA,CTA | P320L | XP_005247980.1 |
XM_011513340.2 | 1247 | Missense Mutation | CCA,CTA | P320L | XP_011511642.1 |
XM_017007583.1 | 1247 | Missense Mutation | CCA,CTA | P320L | XP_016863072.1 |