Product Details

SNP ID
rs115345414
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:153165731 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCTGATCTCACTTAAGTTTGATA[C/T]CAAACACAATTGGGAGGCAATAGTA
Phenotype
MIM: 179541
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAP2B PubMed Links

Gene Details

Gene
RAP2B
Gene Name
RAP2B, member of RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002886.3 3520 UTR 3 NP_002877.2

View Full Product Details