Product Details

SNP ID
rs116055240
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:100609901 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAACCTTAGGGTGCTGGTGGCTGT[C/T]GTGTGTGGACTACTGACTGGCATCA
Phenotype
MIM: 612307
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADGRG7 PubMed Links

Gene Details

Gene
ADGRG7
Gene Name
adhesion G protein-coupled receptor G7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308362.1 301 Intron NP_001295291.1
NM_032787.2 301 Silent Mutation GTC,GTT V15V NP_116176.2
XM_011513245.2 301 UTR 5 XP_011511547.1

View Full Product Details