Product Details

SNP ID
rs115668526
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:167194002 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGGGGAAGTTGAAGAGAGGTTGG[G/T]AATGGGTACAAACATAAAGTTAGAT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZBBX PubMed Links

Gene Details

Gene
ZBBX
Gene Name
zinc finger B-box domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199201.1 Intron NP_001186130.1
NM_001199202.1 Intron NP_001186131.1
NM_024687.3 Intron NP_078963.2
XM_011513144.1 Intron XP_011511446.1
XM_011513145.2 Intron XP_011511447.2
XM_011513148.2 Intron XP_011511450.2
XM_011513149.2 Intron XP_011511451.2
XM_011513151.2 Intron XP_011511453.2
XM_011513152.2 Intron XP_011511454.2
XM_011513153.2 Intron XP_011511455.2
XM_011513155.1 Intron XP_011511457.1
XM_011513156.1 Intron XP_011511458.1
XM_017007186.1 Intron XP_016862675.1
XM_017007187.1 Intron XP_016862676.1
XM_017007188.1 Intron XP_016862677.1
XM_017007189.1 Intron XP_016862678.1
XM_017007190.1 Intron XP_016862679.1

View Full Product Details