Product Details

SNP ID
rs113487931
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:169765003 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAGCACGGCGCCTACGCCCTTCT[C/T]AGTTAGGGTTAGACAAAAAATGGCC
Phenotype
MIM: 608534 MIM: 606042 MIM: 602322
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACTRT3 PubMed Links

Gene Details

Gene
ACTRT3
Gene Name
actin related protein T3
There are no transcripts associated with this gene.

Gene
MYNN
Gene Name
myoneurin
There are no transcripts associated with this gene.

Gene
TERC
Gene Name
telomerase RNA component
There are no transcripts associated with this gene.

View Full Product Details