Product Details

SNP ID
rs116563474
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:171843449 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGGGGATCCTCAGTGTATGCTC[C/T]GGAGTTATTGCTTTCTTTCCTGTCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM212 PubMed Links

Gene Details

Gene
TMEM212
Gene Name
transmembrane protein 212
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164436.1 101 Silent Mutation TCC,TCT S22S NP_001157908.1
XM_011512817.1 101 Silent Mutation TCC,TCT S22S XP_011511119.1
XM_017006376.1 101 Silent Mutation TCC,TCT S22S XP_016861865.1

View Full Product Details