Product Details

SNP ID
rs104893808
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:181712931 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACCCGGGCCTCAATGCGCACGGC[A/G]CAGCGCAGATGCAGCCCATGCACCG
Phenotype
MIM: 184429 MIM: 616338
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SOX2 PubMed Links

Gene Details

Gene
SOX2
Gene Name
SRY-box 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003106.3 1008 Missense Mutation ACA,GCA T191A NP_003097.1
Gene
SOX2-OT
Gene Name
SOX2 overlapping transcript
There are no transcripts associated with this gene.

View Full Product Details