Product Details

SNP ID
rs112889179
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:184164265 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTACTCACTCAGTTTCTCCCTTTC[A/G]GCTGGTGTCAGCTGAGGGCTCACAC
Phenotype
MIM: 601368
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DVL3 PubMed Links

Gene Details

Gene
DVL3
Gene Name
dishevelled segment polarity protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004423.3 Intron NP_004414.3
XM_005247172.2 Intron XP_005247229.1
XM_011512513.2 Intron XP_011510815.1

View Full Product Details