Product Details

SNP ID
rs113181701
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:187722435 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGATTCTGAGAAGGGGCTGGAGA[C/T]GAAAGCATCAACACTCCATGCTTCA
Phenotype
MIM: 109565 MIM: 609138
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BCL6 PubMed Links

Gene Details

Gene
BCL6
Gene Name
B-cell CLL/lymphoma 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130845.1 2509 UTR 3 NP_001124317.1
NM_001134738.1 2509 UTR 3 NP_001128210.1
NM_001706.4 2509 UTR 3 NP_001697.2
XM_005247694.3 2509 UTR 3 XP_005247751.1
XM_011513062.2 2509 UTR 3 XP_011511364.1
Gene
LOC100131635
Gene Name
hCG1645011-like
There are no transcripts associated with this gene.

Gene
RTP2
Gene Name
receptor transporter protein 2
There are no transcripts associated with this gene.

View Full Product Details