Product Details
- SNP ID
-
rs115057120
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:7100024 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGTTGCAGACATTTCCTATAAGTG[G/T]AATCATATATGATCTTTTGTGTATA
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FLJ36777
PubMed Links
Gene Details
- Gene
- FLJ36777
- Gene Name
- uncharacterized LOC730971
There are no transcripts associated with this gene.
View Full Product Details