Product Details

SNP ID
rs114368415
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:2470087 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTCACTTGTCAGTTCCCATAGA[C/T]GTTCACAAACTTAATAATCTTCGTT
Phenotype
MIM: 602850
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CFAP99 PubMed Links

Gene Details

Gene
CFAP99
Gene Name
cilia and flagella associated protein 99
There are no transcripts associated with this gene.

Gene
RNF4
Gene Name
ring finger protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185009.2 Intron NP_001171938.1
NM_001185010.2 Intron NP_001171939.1
NM_002938.4 Intron NP_002929.1

View Full Product Details