Product Details
- SNP ID
-
rs115003408
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:41942406 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAGGGTTCTTCTAATACTTGGATAT[A/G]GGTTGGTGTCTTGCTTGTCAATTGG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM33
PubMed Links
Gene Details
- Gene
- TMEM33
- Gene Name
- transmembrane protein 33
View Full Product Details