Product Details

SNP ID
rs115956256
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:44683303 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAGAAAGTGTTTGATATTCCAAG[C/T]GATGAATGTATTAAGGTAAAATACG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GUF1 PubMed Links

Gene Details

Gene
GUF1
Gene Name
GUF1 homolog, GTPase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021927.2 715 Silent Mutation AGC,AGT S218S NP_068746.2
XM_005248122.3 715 UTR 5 XP_005248179.1
XM_011513732.2 715 Silent Mutation AGC,AGT S218S XP_011512034.1

View Full Product Details