Product Details

SNP ID
rs115212906
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:48498783 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAGACGTTATCACCAAACATCCA[C/T]ACCTTGAAAATATGACTATCACATA
Phenotype
MIM: 607520
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FRYL PubMed Links

Gene Details

Gene
FRYL
Gene Name
FRY like transcription coactivator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015030.1 6977 UTR 3 NP_055845.1
XM_005248090.4 6977 UTR 3 XP_005248147.1
XM_005248093.4 6977 UTR 3 XP_005248150.1
XM_011513680.2 6977 UTR 3 XP_011511982.1
XM_011513681.2 6977 UTR 3 XP_011511983.1
XM_011513682.2 6977 UTR 3 XP_011511984.1
XM_011513683.2 6977 UTR 3 XP_011511985.1
XM_011513684.2 6977 UTR 3 XP_011511986.1
XM_011513685.2 6977 Intron XP_011511987.1
XM_017008046.1 6977 UTR 3 XP_016863535.1
Gene
SLC10A4
Gene Name
solute carrier family 10 member 4
There are no transcripts associated with this gene.

Gene
ZAR1
Gene Name
zygote arrest 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175619.2 6977 Intron NP_783318.1

View Full Product Details