Product Details

SNP ID
rs114109706
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:25662759 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGAACTTCTGCCGTCCTACTCCA[C/T]GGCTACACTGATAGATGAGCCCACT
Phenotype
MIM: 604217
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC34A2 PubMed Links

Gene Details

Gene
SLC34A2
Gene Name
solute carrier family 34 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177998.1 268 Missense Mutation ACG,ATG T55M NP_001171469.1
NM_001177999.1 268 Missense Mutation ACG,ATG T55M NP_001171470.1
NM_006424.2 268 Missense Mutation ACG,ATG T56M NP_006415.2

View Full Product Details