Product Details
- SNP ID
-
rs116150154
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:15002875 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAGGCCGCCTCCCCCTTCTCCGTC[C/T]CCCTCGGCGGCGGCGCGGGCAGCCC
- Phenotype
-
MIM: 610605
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CPEB2
PubMed Links
Gene Details
- Gene
- CPEB2
- Gene Name
- cytoplasmic polyadenylation element binding protein 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001177381.1 |
425 |
Missense Mutation |
CCC,TCC |
P68S |
NP_001170852.1 |
NM_001177382.1 |
425 |
Missense Mutation |
CCC,TCC |
P68S |
NP_001170853.1 |
NM_001177383.1 |
425 |
Missense Mutation |
CCC,TCC |
P68S |
NP_001170854.1 |
NM_001177384.1 |
425 |
Missense Mutation |
CCC,TCC |
P68S |
NP_001170855.1 |
NM_182485.2 |
425 |
Missense Mutation |
CCC,TCC |
P68S |
NP_872291.2 |
NM_182646.2 |
425 |
Missense Mutation |
CCC,TCC |
P68S |
NP_872587.2 |
XM_005248135.3 |
425 |
Missense Mutation |
CCC,TCC |
P68S |
XP_005248192.2 |
XM_011513777.2 |
425 |
Missense Mutation |
CCC,TCC |
P68S |
XP_011512079.1 |
XM_011513778.2 |
425 |
Missense Mutation |
CCC,TCC |
P68S |
XP_011512080.1 |
XM_011513779.1 |
425 |
Missense Mutation |
CCC,TCC |
P68S |
XP_011512081.1 |
XM_017007733.1 |
425 |
Intron |
|
|
XP_016863222.1 |
XM_017007734.1 |
425 |
Intron |
|
|
XP_016863223.1 |
- Gene
- CPEB2-AS1
- Gene Name
- CPEB2 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.
- Gene
- LOC101929095
- Gene Name
- uncharacterized LOC101929095
There are no transcripts associated with this gene.
View Full Product Details