Product Details

SNP ID
rs116230084
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:17634080 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCATTTTGTATTATAAATAAATAT[A/G]TATGTTCACAACCTCTTAACCAAAA
Phenotype
MIM: 610311
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM184B PubMed Links

Gene Details

Gene
FAM184B
Gene Name
family with sequence similarity 184 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015688.1 Intron NP_056503.1
Gene
MED28
Gene Name
mediator complex subunit 28
There are no transcripts associated with this gene.

View Full Product Details