Product Details

SNP ID
rs115081846
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:10280327 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCTACTGAATTTGTGTTTCAAAT[G/T]AAATAATCAATTTTAGGATTCCTAC
Phenotype
MIM: 613379
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CMBL PubMed Links

Gene Details

Gene
CMBL
Gene Name
carboxymethylenebutenolidase homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138809.3 1315 UTR 3 NP_620164.1
XM_017009034.1 1315 UTR 3 XP_016864523.1
XM_017009035.1 1315 UTR 3 XP_016864524.1

View Full Product Details