Product Details

SNP ID
rs116033240
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:14673760 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTATTTATAGAGTTCTTATTGTTT[A/G]TAGCAGAAAATGCAGTTAAATCCGT
Phenotype
MIM: 615712
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OTULIN PubMed Links

Gene Details

Gene
OTULIN
Gene Name
OTU deubiquitinase with linear linkage specificity
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138348.4 Intron NP_612357.4
XM_011514151.2 Intron XP_011512453.1
XM_011514152.2 Intron XP_011512454.1
XM_011514154.2 Intron XP_011512456.1
XM_017010015.1 Intron XP_016865504.1

View Full Product Details