Product Details
- SNP ID
-
rs115200817
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:1708787 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGGGCAGCTTTGTCTAGACATGGA[A/G]GGGCACAAGGACTTGAGCTGAGCAG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR4277
PubMed Links
Gene Details
- Gene
- MIR4277
- Gene Name
- microRNA 4277
There are no transcripts associated with this gene.
View Full Product Details