Product Details
- SNP ID
-
rs115866543
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:21575944 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTTGTGTATGCATAAGCTACAACC[C/T]GTTGAAGACAAAAGAAAGTGTTGAG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GUSBP1
PubMed Links
Gene Details
- Gene
- GUSBP1
- Gene Name
- glucuronidase, beta pseudogene 1
There are no transcripts associated with this gene.
View Full Product Details