Product Details

SNP ID
rs116547736
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:32533349 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGTATTGGTGGCAAGTCATCTTG[G/T]ACCATGAGGATGAGGACAACCCCCC
Phenotype
MIM: 600503
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SUB1 PubMed Links

Gene Details

Gene
SUB1
Gene Name
SUB1 homolog, transcriptional regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006713.3 Intron NP_006704.3
XM_011513944.2 Intron XP_011512246.1
XM_017008986.1 Intron XP_016864475.1
XM_017008987.1 Intron XP_016864476.1

View Full Product Details