Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001161429.2 | Intron | NP_001154901.1 | |||
NM_001323273.1 | Intron | NP_001310202.1 | |||
NM_001323274.1 | Intron | NP_001310203.1 | |||
NM_001323275.1 | Intron | NP_001310204.1 | |||
NM_001323276.1 | Intron | NP_001310205.1 | |||
NM_001323277.1 | Intron | NP_001310206.1 | |||
NM_001323278.1 | Intron | NP_001310207.1 | |||
NM_001323279.1 | Intron | NP_001310208.1 | |||
NM_001323280.1 | Intron | NP_001310209.1 | |||
NM_145000.4 | Intron | NP_659437.3 | |||
XM_006714452.3 | Intron | XP_006714515.1 | |||
XM_006714453.3 | Intron | XP_006714516.1 | |||
XM_006714455.3 | Intron | XP_006714518.1 |