Product Details

SNP ID
rs114306678
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:73632241 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGAGGGCAAAGTTTGCTCAGGAA[C/T]TAGACAGTTAGTCCTTCCCAGATGT
Phenotype
MIM: 612790
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGEF28 PubMed Links

Gene Details

Gene
ARHGEF28
Gene Name
Rho guanine nucleotide exchange factor 28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080479.2 Intron NP_001073948.2
NM_001177693.1 Intron NP_001171164.1
NM_001244364.1 Intron NP_001231293.1
XM_005248568.4 Intron XP_005248625.1
XM_011543546.1 Intron XP_011541848.1
XM_011543547.2 Intron XP_011541849.1
XM_017009702.1 Intron XP_016865191.1
XM_017009703.1 Intron XP_016865192.1

View Full Product Details