Product Details

SNP ID
rs114759904
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:87313259 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACAGGTGGTCTTTAGCTAATCTA[A/G]TATCTTTGAGGTGGCTGAATTTTGG
Phenotype
MIM: 601953 MIM: 139150
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCNH PubMed Links

Gene Details

Gene
CCNH
Gene Name
cyclin H
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199189.1 Intron NP_001186118.1
NM_001239.3 Intron NP_001230.1
XM_005248627.3 Intron XP_005248684.1
XM_005248629.4 Intron XP_005248686.1
XM_011543706.2 Intron XP_011542008.1
XM_017010014.1 Intron XP_016865503.1
Gene
LOC644285
Gene Name
uncharacterized LOC644285
There are no transcripts associated with this gene.

Gene
RASA1
Gene Name
RAS p21 protein activator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002890.2 Intron NP_002881.1
NM_022650.2 Intron NP_072179.1
XM_011543525.1 Intron XP_011541827.1
XM_011543527.2 Intron XP_011541829.1

View Full Product Details