Product Details

SNP ID
rs112026794
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:54518072 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGGACATCGAGGGCTGGCTCGA[A/G]GGGGTCAACAGCCGCGGCGACCGCG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SNX18 PubMed Links

Gene Details

Gene
SNX18
Gene Name
sorting nexin 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102575.1 310 Silent Mutation GAA,GAG E40E NP_001096045.1
NM_001145427.1 310 Silent Mutation GAA,GAG E40E NP_001138899.1
NM_052870.2 310 Silent Mutation GAA,GAG E40E NP_443102.2
XM_017008997.1 310 Silent Mutation GAA,GAG E40E XP_016864486.1

View Full Product Details