Product Details

SNP ID
rs111518577
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:55160344 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATCAACAGCTTTTATGCCTTTG[A/G]AGTGAAGGATGCAAAAGGAAGAACT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDC20B PubMed Links

Gene Details

Gene
CDC20B
Gene Name
cell division cycle 20B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145734.2 227 Intron NP_001139206.2
NM_001170402.1 227 Intron NP_001163873.1
NM_152623.2 227 Intron NP_689836.2
XM_011543218.2 227 Intron XP_011541520.1
Gene
GPX8
Gene Name
glutathione peroxidase 8 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008397.3 227 Missense Mutation GAA,GGA E51G NP_001008398.2
NM_001306197.1 227 Intron NP_001293126.1
NM_001306198.1 227 Missense Mutation GAA,GGA E51G NP_001293127.1
NM_001306201.1 227 Intron NP_001293130.1
XM_006714631.2 227 Missense Mutation GAA,GGA E51G XP_006714694.1

View Full Product Details