Product Details

SNP ID
rs116183617
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:73354144 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGCTTTCAGCAGCCGCGTCTGGA[C/T]CTGGAACACCTCTGGATCTTTCAGG
Phenotype
MIM: 611111 MIM: 611687
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DPPA5 PubMed Links

Gene Details

Gene
DPPA5
Gene Name
developmental pluripotency associated 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025290.2 Intron NP_001020461.1
Gene
KHDC3L
Gene Name
KH domain containing 3 like, subcortical maternal complex member
There are no transcripts associated with this gene.

View Full Product Details