Product Details

SNP ID
rs113430068
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:75237771 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTTGAAATTATTGGCATAAAGTG[A/G]TAAAATCAGACAAAGCATGGTAAAG
Phenotype
MIM: 123996
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COX7A2 PubMed Links

Gene Details

Gene
COX7A2
Gene Name
cytochrome c oxidase subunit 7A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001865.3 605 UTR 3 NP_001856.2

View Full Product Details