Product Details

SNP ID
rs116588022
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:78867577 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTATGGAAACCCAGCTGCCATCGCT[A/G]TGTCTCTGCAAAAGACCCCTCCGAC
Phenotype
MIM: 615375
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IRAK1BP1 PubMed Links

Gene Details

Gene
IRAK1BP1
Gene Name
interleukin 1 receptor associated kinase 1 binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010844.3 92 Missense Mutation ATG,GTG M1V NP_001010844.1
XM_006715336.3 92 Missense Mutation ATG,GTG M1V XP_006715399.1
XM_011535447.1 92 Missense Mutation ATG,GTG M1V XP_011533749.1
XM_011535448.2 92 Missense Mutation ATG,GTG M1V XP_011533750.1
XM_017010264.1 92 Intron XP_016865753.1
XM_017010265.1 92 Missense Mutation ATG,GTG M1V XP_016865754.1

View Full Product Details