Product Details
- SNP ID
-
rs116875076
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:83512979 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGGGACCCTCTTTATTTTTATGAT[G/T]GTTAACAGATGCCATTGAGAAAGGT
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PRSS35
PubMed Links
Gene Details
- Gene
- PRSS35
- Gene Name
- protease, serine 35
View Full Product Details