Product Details

SNP ID
rs114081526
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:85452283 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTGTGACACTCTTTTCTCCACA[A/G]TTGTGTGAACCCTTTTTAGTGGAAA
Phenotype
MIM: 129190
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NT5E PubMed Links

Gene Details

Gene
NT5E
Gene Name
5'-nucleotidase ecto
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204813.1 Intron NP_001191742.1
NM_002526.3 Intron NP_002517.1

View Full Product Details