Product Details

SNP ID
rs116706541
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:61557400 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATAGAATATATATAGAGAGTGTCT[C/T]TCCTGCCATGTCAGACAGAAGGCTG
Phenotype
MIM: 610487
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KHDRBS2 PubMed Links

Gene Details

Gene
KHDRBS2
Gene Name
KH RNA binding domain containing, signal transduction associated 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152688.3 Intron NP_689901.2
XM_011535515.2 Intron XP_011533817.1
XM_011535516.1 Intron XP_011533818.1
XM_011535517.1 Intron XP_011533819.1
XM_011535519.1 Intron XP_011533821.1
XM_011535520.1 Intron XP_011533822.1
XM_017010334.1 Intron XP_016865823.1
XM_017010335.1 Intron XP_016865824.1
XM_017010336.1 Intron XP_016865825.1
XM_017010337.1 Intron XP_016865826.1
XM_017010338.1 Intron XP_016865827.1
XM_017010339.1 Intron XP_016865828.1
XM_017010340.1 Intron XP_016865829.1
XM_017010341.1 Intron XP_016865830.1
XM_017010342.1 Intron XP_016865831.1
XM_017010343.1 Intron XP_016865832.1
XM_017010344.1 Intron XP_016865833.1
XM_017010345.1 Intron XP_016865834.1
XM_017010346.1 Intron XP_016865835.1
XM_017010347.1 Intron XP_016865836.1
XM_017010348.1 Intron XP_016865837.1

View Full Product Details