Product Details

SNP ID
rs118078855
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:70568784 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAATTTTCACTTCTACCAGCAGT[A/G]TATTGGAGTTCTGTTGTTCCACAAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM135A PubMed Links

Gene Details

Gene
FAM135A
Gene Name
family with sequence similarity 135 member A
There are no transcripts associated with this gene.

Gene
SDHAF4
Gene Name
succinate dehydrogenase complex assembly factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145267.2 Intron NP_660310.2

View Full Product Details