Product Details
- SNP ID
-
rs114752296
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:26597156 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGGCCATATCCCGCCGCGCTTCCG[A/G]CCCCTGCACGTCCGCAACCTTCTCA
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ABT1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3800302] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ABT1
- Gene Name
- activator of basal transcription 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_013375.3 |
214 |
Silent Mutation |
CGA,CGG |
R58R |
NP_037507.1 |
- Gene
- LOC105374989
- Gene Name
- uncharacterized LOC105374989
There are no transcripts associated with this gene.
View Full Product Details