Product Details

SNP ID
rs112785172
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:27893099 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGGAGCCCAGCTCTAGAGGAGCG[A/G]GTTTTGGCCTTGGCGCGAGCCTTGC
Phenotype
MIM: 602796 MIM: 602808 MIM: 602817
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HIST1H2AM PubMed Links

Gene Details

Gene
HIST1H2AM
Gene Name
histone cluster 1, H2am
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003514.2 Intron NP_003505.1
Gene
HIST1H2BO
Gene Name
histone cluster 1, H2bo
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003527.4 Intron NP_003518.2
Gene
HIST1H3J
Gene Name
histone cluster 1, H3j
There are no transcripts associated with this gene.

View Full Product Details