Product Details

SNP ID
rs117540842
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:29723857 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGGGAGTGGACCACAGGGTACGC[C/G]AAGGCCAACGCACAGACTGACCGAG
Phenotype
MIM: 143110
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HLA-F PubMed Links

Gene Details

Gene
HLA-F
Gene Name
major histocompatibility complex, class I, F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098478.1 294 Silent Mutation GCC,GCG A88A NP_001091948.1
NM_001098479.1 294 Silent Mutation GCC,GCG A88A NP_001091949.1
NM_018950.2 294 Silent Mutation GCC,GCG A88A NP_061823.2
XM_011514564.1 294 Silent Mutation GCC,GCG A88A XP_011512866.1
XM_017010810.1 294 Silent Mutation GCC,GCG A88A XP_016866299.1
XM_017010811.1 294 Silent Mutation GCC,GCG A88A XP_016866300.1
XM_017010812.1 294 Silent Mutation GCC,GCG A88A XP_016866301.1
XM_017010813.1 294 Silent Mutation GCC,GCG A88A XP_016866302.1
XM_017010814.1 294 UTR 5 XP_016866303.1
XM_017010815.1 294 UTR 5 XP_016866304.1
Gene
HLA-F-AS1
Gene Name
HLA-F antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details